Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS10
Entrez Gene ID
55174
Gene Name
integrator complex subunit 10
Gene Aliases
C8orf35, INT10
Chromosome Location
Chr.8: 19817149 - 19852078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018142.2 NP_060612.2 5 - 6 651 122
XM_005273555.3 XP_005273612.1 5 - 6 909 122
XM_005273556.3 XP_005273613.1 5 - 6 693 122
XM_005273557.3 XP_005273614.1 5 - 6 693 122
XM_006716355.2 XP_006716418.1 5 - 6 798 122
XM_017013606.1 XP_016869095.1 5 - 6 693 122
XM_017013607.1 XP_016869096.1 5 - 6 909 122
XM_017013608.1 XP_016869097.1 5 - 6 474 122
XM_017013609.1 XP_016869098.1 5 - 6 798 122
GenBank mRNA AK001431.1 - 5 - 6 571 122
AK092931.1 - 1 - 2 1569 122
BC001915.1 - 3 - 4 288 122
BC006209.2 - 3 - 4 288 122
BC051282.1 - 3 - 4 302 122
CR457262.1 - 2 - 3 181 122

View Full Product Details