The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018142.2 | NP_060612.2 | 9 - 10 | 1274 | 67 |
XM_005273556.3 | XP_005273613.1 | 9 - 10 | 1316 | 67 | |
XM_017013608.1 | XP_016869097.1 | 8 - 9 | 927 | 67 | |
XM_017013609.1 | XP_016869098.1 | 8 - 9 | 1251 | 67 | |
GenBank mRNA | AK001431.1 | - | 9 - 10 | 1194 | 67 |
BC001915.1 | - | 7 - 8 | 911 | 67 | |
BC006209.2 | - | 7 - 8 | 911 | 67 | |
BC051282.1 | - | 7 - 8 | 925 | 67 | |
CR457262.1 | - | 6 - 7 | 804 | 67 |