The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004106.1 | NP_004097.1 | 2 - 3 | 171 | 89 |
GenBank mRNA | AK307102.1 | - | 2 - 3 | 196 | 89 |
M33195.1 | - | 2 - 3 | 171 | 89 |