The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142777.1 | NP_001136249.1 | 3 - 4 | 621 | 63 |
NM_001142778.1 | NP_001136250.1 | 3 - 4 | 428 | 63 | |
NM_025193.3 | NP_079469.2 | 3 - 4 | 428 | 63 | |
XM_005255601.3 | XP_005255658.2 | 3 - 4 | 409 | 63 | |
XM_011545960.2 | XP_011544262.1 | 3 - 4 | 606 | 63 | |
XM_011545961.1 | XP_011544263.1 | 3 - 4 | 612 | 63 | |
XM_011545962.2 | XP_011544264.1 | 3 - 4 | 409 | 63 | |
XM_017023732.1 | XP_016879221.1 | 3 - 4 | 590 | 63 | |
GenBank mRNA | AF277719.1 | - | 2 - 3 | 340 | 63 |
AK057436.1 | - | 3 - 4 | 612 | 63 | |
AK290950.1 | - | 3 - 4 | 382 | 63 | |
AK292068.1 | - | 3 - 4 | 572 | 63 | |
BC004929.1 | - | 3 - 4 | 415 | 63 |