The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014712.2 | NP_055527.1 | 17 - 18 | 5501 | 66 |
XM_005255723.1 | XP_005255780.1 | 17 - 18 | 4985 | 66 | |
XM_006721106.3 | XP_006721169.1 | 17 - 18 | 4906 | 66 | |
XM_017023909.1 | XP_016879398.1 | 17 - 18 | 6138 | 66 | |
GenBank mRNA | AB002337.2 | - | 17 - 18 | 5501 | 66 |
BC027450.1 | - | 4 - 5 | 1101 | 66 |