Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SETD1A
Entrez Gene ID
9739
Gene Name
SET domain containing 1A
Gene Aliases
KMT2F, Set1, Set1A
Chromosome Location
Chr.16: 30956618 - 30984664 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014712.2 NP_055527.1 10 - 11 3462 141
XM_005255723.1 XP_005255780.1 10 - 11 2946 141
XM_006721106.3 XP_006721169.1 10 - 11 2867 141
XM_017023909.1 XP_016879398.1 10 - 11 4099 141
GenBank mRNA AB002337.2 - 10 - 11 3462 141

View Full Product Details