Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRNAD1
Entrez Gene ID
51093
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Gene Aliases
C1orf66, CGI-41, METTL25B
Chromosome Location
Chr.1: 156728471 - 156736960 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015997.3 NP_057081.3 6 - 7 1758 61
XM_005245234.4 XP_005245291.1 6 - 7 1543 61
XM_005245235.4 XP_005245292.1 5 - 6 1402 61
XM_011509600.1 XP_011507902.1 6 - 7 1095 61
XM_017001404.1 XP_016856893.1 6 - 7 1678 61
XM_017001405.1 XP_016856894.1 6 - 7 1623 61
XM_017001406.1 XP_016856895.1 6 - 7 1629 61
GenBank mRNA AF151800.1 - 6 - 7 1348 61
AK000412.1 - 7 - 8 1574 61
BC011382.1 - 6 - 7 1743 61

View Full Product Details