The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005599.3 | NP_005590.1 | 1 - 2 | 174 | 79 |
XM_017001370.1 | XP_016856859.1 | 1 - 2 | 174 | 79 | |
GenBank mRNA | BC096359.3 | - | 1 - 2 | 120 | 79 |
BC096360.1 | - | 1 - 2 | 120 | 79 | |
DA608185.1 | - | 1 - 2 | 174 | 79 | |
M96740.1 | - | 1 - 2 | 173 | 79 |