The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190326.1 | NP_001177255.1 | 8 - 9 | 2110 | 97 |
NM_022720.6 | NP_073557.3 | 9 - 10 | 2209 | 97 | |
XM_006724268.2 | XP_006724331.1 | 6 - 7 | 844 | 97 | |
GenBank mRNA | AK025539.1 | - | 8 - 9 | 1173 | 97 |
AK025780.1 | - | 4 - 5 | 485 | 97 | |
AK222796.1 | - | 8 - 9 | 1173 | 97 | |
AY189282.1 | - | 5 - 6 | 503 | 97 | |
BC009323.2 | - | 7 - 8 | 1012 | 97 | |
BC037564.1 | - | 9 - 10 | 2159 | 97 | |
BC078147.1 | - | 7 - 8 | 1824 | 97 | |
BX649187.1 | - | 6 - 7 | 1101 | 97 | |
CR456356.1 | - | 8 - 9 | 1867 | 97 |