The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267045.1 | NP_001253974.1 | 5 - 6 | 957 | 101 |
NM_005789.3 | NP_005780.2 | 3 - 4 | 630 | 101 | |
NM_176863.2 | NP_789839.1 | 3 - 4 | 630 | 101 | |
NR_049772.1 | 3 - 4 | 630 | 101 | ||
XM_011524177.2 | XP_011522479.1 | 3 - 4 | 639 | 101 | |
GenBank mRNA | AK057221.1 | - | 5 - 6 | 796 | 101 |
AK074999.1 | - | 3 - 4 | 327 | 101 | |
AK292618.1 | - | 3 - 4 | 355 | 101 | |
AK303211.1 | - | 3 - 4 | 327 | 101 | |
AK307110.1 | - | 2 - 3 | 1031 | 101 | |
AK314347.1 | - | 3 - 4 | 373 | 101 | |
BC001423.2 | - | 3 - 4 | 357 | 101 | |
BC002684.1 | - | 3 - 4 | 336 | 101 | |
BC008020.1 | - | 3 - 4 | 288 | 101 | |
BI464061.1 | - | 3 - 4 | 633 | 101 | |
BT019386.1 | - | 3 - 4 | 140 | 101 | |
BX640801.1 | - | 5 - 6 | 684 | 102 | |
EU794653.1 | - | 3 - 4 | 357 | 101 | |
FJ200488.1 | - | 3 - 4 | 140 | 101 | |
U11292.1 | - | 3 - 4 | 302 | 101 |