Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNPEPL1
Entrez Gene ID
57140
Gene Name
arginyl aminopeptidase like 1
Gene Aliases
-
Chromosome Location
Chr.2: 240568526 - 240578732 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018226.4 NP_060696.4 7 - 8 1400 100
XM_005247036.4 XP_005247093.1 7 - 8 1461 100
GenBank mRNA AF300795.1 - 7 - 8 1106 100
AK001668.1 - 3 - 4 364 100
BC017301.2 - 4 - 5 552 100
BC067258.1 - 7 - 8 975 100
BC082975.1 - 7 - 8 1300 100
BC099906.1 - 5 - 6 1513 100

View Full Product Details