Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNPEPL1
Entrez Gene ID
57140
Gene Name
arginyl aminopeptidase like 1
Gene Aliases
-
Chromosome Location
Chr.2: 240568526 - 240578732 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005247036.4 XP_005247093.1 8 - 9 1574 88
GenBank mRNA AK130185.1 - 1 - 2 114 88

View Full Product Details