Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF1A
Entrez Gene ID
547
Gene Name
kinesin family member 1A
Gene Aliases
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
Chromosome Location
Chr.2: 240713764 - 240821025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244008.1 NP_001230937.1 22 - 23 2270 64
NM_001320705.1 NP_001307634.1 22 - 23 2270 64
NM_004321.6 NP_004312.2 20 - 21 2056 64
XM_005247022.1 XP_005247079.1 22 - 23 2181 64
XM_005247024.1 XP_005247081.1 21 - 22 2154 64
XM_005247027.1 XP_005247084.1 21 - 22 2154 64
XM_005247028.2 XP_005247085.1 21 - 22 2157 64
XM_006712605.1 XP_006712668.1 21 - 22 2154 64
XM_011511364.1 XP_011509666.1 22 - 23 2193 64
XM_011511365.1 XP_011509667.1 22 - 23 2181 64
XM_011511366.2 XP_011509668.1 12 - 13 1253 64
XM_011511367.1 XP_011509669.1 12 - 13 1183 64
XM_017004383.1 XP_016859872.1 22 - 23 2256 64
XM_017004384.1 XP_016859873.1 22 - 23 2268 64
XM_017004385.1 XP_016859874.1 22 - 23 2256 64
XM_017004386.1 XP_016859875.1 22 - 23 2256 64
XM_017004387.1 XP_016859876.1 22 - 23 2256 64
XM_017004388.1 XP_016859877.1 22 - 23 2256 64
XM_017004389.1 XP_016859878.1 22 - 23 2256 64
XM_017004390.1 XP_016859879.1 21 - 22 2154 64
XM_017004391.1 XP_016859880.1 21 - 22 2035 64
GenBank mRNA AB210029.1 - 22 - 23 2270 64
AB290172.1 - 22 - 23 2270 64
BC111780.1 - 20 - 21 2024 64
BC111799.1 - 20 - 21 2024 64
BF879579.1 - 4 - 5 446 64
KJ534875.1 - 20 - 21 1996 64

View Full Product Details