Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF1A
Entrez Gene ID
547
Gene Name
kinesin family member 1A
Gene Aliases
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
Chromosome Location
Chr.2: 240713764 - 240821025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244008.1 NP_001230937.1 3 - 4 428 63
NM_001320705.1 NP_001307634.1 3 - 4 428 63
NM_004321.6 NP_004312.2 2 - 3 241 63
XM_005247022.1 XP_005247079.1 3 - 4 339 63
XM_005247024.1 XP_005247081.1 3 - 4 339 63
XM_005247027.1 XP_005247084.1 3 - 4 339 63
XM_005247028.2 XP_005247085.1 3 - 4 342 63
XM_006712605.1 XP_006712668.1 3 - 4 339 63
XM_011511364.1 XP_011509666.1 3 - 4 351 63
XM_011511365.1 XP_011509667.1 3 - 4 339 63
XM_017004383.1 XP_016859872.1 3 - 4 339 63
XM_017004384.1 XP_016859873.1 3 - 4 351 63
XM_017004385.1 XP_016859874.1 3 - 4 339 63
XM_017004386.1 XP_016859875.1 3 - 4 339 63
XM_017004387.1 XP_016859876.1 3 - 4 339 63
XM_017004388.1 XP_016859877.1 3 - 4 339 63
XM_017004389.1 XP_016859878.1 3 - 4 339 63
XM_017004390.1 XP_016859879.1 3 - 4 339 63
XM_017004391.1 XP_016859880.1 2 - 3 193 63
GenBank mRNA AB210029.1 - 3 - 4 428 63
AB290172.1 - 3 - 4 428 63
AB621817.1 - 3 - 4 355 63
AI815863.1 - 3 - 4 327 63
AI878864.1 - 3 - 4 327 63
BC064906.1 - 3 - 4 331 63
BC111780.1 - 2 - 3 209 63
BC111799.1 - 2 - 3 209 63
KJ534874.1 - 2 - 3 181 63
KJ534875.1 - 2 - 3 181 63

View Full Product Details