Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF1A
Entrez Gene ID
547
Gene Name
kinesin family member 1A
Gene Aliases
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
Chromosome Location
Chr.2: 240713764 - 240821025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244008.1 NP_001230937.1 43 - 44 4907 107
NM_001320705.1 NP_001307634.1 42 - 43 4631 107
NM_004321.6 NP_004312.2 40 - 41 4417 107
XM_005247022.1 XP_005247079.1 44 - 45 4845 107
XM_005247024.1 XP_005247081.1 43 - 44 4818 107
XM_005247027.1 XP_005247084.1 42 - 43 4539 107
XM_005247028.2 XP_005247085.1 41 - 42 4518 107
XM_006712605.1 XP_006712668.1 42 - 43 4791 107
XM_011511364.1 XP_011509666.1 44 - 45 4857 107
XM_011511365.1 XP_011509667.1 43 - 44 4569 107
XM_011511366.2 XP_011509668.1 34 - 35 3917 107
XM_011511367.1 XP_011509669.1 34 - 35 3847 107
XM_017004383.1 XP_016859872.1 44 - 45 4920 107
XM_017004384.1 XP_016859873.1 44 - 45 4932 107
XM_017004385.1 XP_016859874.1 44 - 45 4917 107
XM_017004386.1 XP_016859875.1 43 - 44 4893 107
XM_017004387.1 XP_016859876.1 43 - 44 4644 107
XM_017004388.1 XP_016859877.1 43 - 44 4641 107
XM_017004389.1 XP_016859878.1 42 - 43 4617 107
XM_017004390.1 XP_016859879.1 42 - 43 4542 107
XM_017004391.1 XP_016859880.1 43 - 44 4696 107
GenBank mRNA AB209138.1 - 16 - 17 1716 107
AB290172.1 - 43 - 44 4907 107
AF038173.1 - 4 - 5 544 107
AF052089.1 - 4 - 5 544 107
AK130880.1 - 4 - 5 1132 107
BC111780.1 - 40 - 41 4385 107
BC111799.1 - 40 - 41 4385 107
BX537556.1 - 3 - 4 1219 107
KJ534875.1 - 40 - 41 4357 107
KJ535053.1 - 5 - 6 560 107
L79946.1 - 2 - 3 348 107
U37194.1 - 12 - 13 1270 107

View Full Product Details