Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF1A
Entrez Gene ID
547
Gene Name
kinesin family member 1A
Gene Aliases
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
Chromosome Location
Chr.2: 240713764 - 240821025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244008.1 NP_001230937.1 9 - 10 1114 91
NM_001320705.1 NP_001307634.1 9 - 10 1114 91
NM_004321.6 NP_004312.2 8 - 9 927 91
XM_005247022.1 XP_005247079.1 9 - 10 1025 91
XM_005247024.1 XP_005247081.1 9 - 10 1025 91
XM_005247027.1 XP_005247084.1 9 - 10 1025 91
XM_005247028.2 XP_005247085.1 9 - 10 1028 91
XM_006712605.1 XP_006712668.1 9 - 10 1025 91
XM_011511364.1 XP_011509666.1 9 - 10 1037 91
XM_011511365.1 XP_011509667.1 9 - 10 1025 91
XM_017004383.1 XP_016859872.1 9 - 10 1025 91
XM_017004384.1 XP_016859873.1 9 - 10 1037 91
XM_017004385.1 XP_016859874.1 9 - 10 1025 91
XM_017004386.1 XP_016859875.1 9 - 10 1025 91
XM_017004387.1 XP_016859876.1 9 - 10 1025 91
XM_017004388.1 XP_016859877.1 9 - 10 1025 91
XM_017004389.1 XP_016859878.1 9 - 10 1025 91
XM_017004390.1 XP_016859879.1 9 - 10 1025 91
XM_017004391.1 XP_016859880.1 8 - 9 879 91
GenBank mRNA AB210029.1 - 9 - 10 1114 91
AB290172.1 - 9 - 10 1114 91
AL706949.1 - 3 - 4 238 91
BC111780.1 - 8 - 9 895 91
BC111799.1 - 8 - 9 895 91
KJ534875.1 - 8 - 9 867 91

View Full Product Details