Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGGY
Entrez Gene ID
55277
Gene Name
FGGY carbohydrate kinase domain containing
Gene Aliases
-
Chromosome Location
Chr.1: 59296405 - 59762730 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113411.1 NP_001106882.1 11 - 12 1431 71
XM_011541730.1 XP_011540032.1 11 - 12 1355 71
XM_011541731.1 XP_011540033.1 11 - 12 1289 71
XM_011541732.1 XP_011540034.1 11 - 12 1332 71
XM_011541733.1 XP_011540035.1 11 - 12 1250 71
XM_011541736.2 XP_011540038.1 9 - 10 1196 71
XM_017001643.1 XP_016857132.1 11 - 12 1461 71
XM_017001644.1 XP_016857133.1 11 - 12 1254 71
XM_017001648.1 XP_016857137.1 12 - 13 1274 71
XM_017001649.1 XP_016857138.1 11 - 12 1400 71
XM_017001654.1 XP_016857143.1 9 - 10 1151 71
XM_017001655.1 XP_016857144.1 9 - 10 1060 71
XM_017001656.1 XP_016857145.1 12 - 13 1467 71
XM_017001657.1 XP_016857146.1 11 - 12 1255 71
XM_017001658.1 XP_016857147.1 12 - 13 1398 71
XM_017001659.1 XP_016857148.1 10 - 11 1288 71
XM_017001660.1 XP_016857149.1 10 - 11 1191 71
XM_017001661.1 XP_016857150.1 10 - 11 1348 71
XM_017001662.1 XP_016857151.1 10 - 11 1033 71
XM_017001663.1 XP_016857152.1 10 - 11 1208 71
XM_017001664.1 XP_016857153.1 9 - 10 1085 71
XM_017001665.1 XP_016857154.1 9 - 10 992 71
XM_017001668.1 XP_016857157.1 9 - 10 956 71
XM_017001669.1 XP_016857158.1 9 - 10 1051 71
XM_017001670.1 XP_016857159.1 9 - 10 1824 71
XM_017001671.1 XP_016857160.1 9 - 10 952 71
XM_017001675.1 XP_016857164.1 10 - 11 1288 71
XM_017001677.1 XP_016857166.1 7 - 8 3526 71
XM_017001678.1 XP_016857167.1 7 - 8 4801 71
GenBank mRNA BX647626.1 - 11 - 12 1403 71

View Full Product Details