Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGGY
Entrez Gene ID
55277
Gene Name
FGGY carbohydrate kinase domain containing
Gene Aliases
-
Chromosome Location
Chr.1: 59296405 - 59762730 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113411.1 NP_001106882.1 3 - 4 521 89
NM_018291.3 NP_060761.3 3 - 4 521 89
NR_103473.1 4 - 5 479 89
XM_011541730.1 XP_011540032.1 3 - 4 445 89
XM_011541731.1 XP_011540033.1 3 - 4 379 89
XM_011541732.1 XP_011540034.1 3 - 4 422 89
XM_011541733.1 XP_011540035.1 3 - 4 340 89
XM_011541735.1 XP_011540037.1 3 - 4 340 89
XM_017001643.1 XP_016857132.1 3 - 4 551 89
XM_017001644.1 XP_016857133.1 3 - 4 344 89
XM_017001645.1 XP_016857134.1 3 - 4 445 89
XM_017001646.1 XP_016857135.1 3 - 4 422 89
XM_017001647.1 XP_016857136.1 3 - 4 550 89
XM_017001648.1 XP_016857137.1 4 - 5 364 89
XM_017001649.1 XP_016857138.1 3 - 4 490 89
XM_017001650.1 XP_016857139.1 5 - 6 713 89
XM_017001651.1 XP_016857140.1 5 - 6 569 89
XM_017001652.1 XP_016857141.1 3 - 4 490 89
XM_017001653.1 XP_016857142.1 3 - 4 422 89
XM_017001656.1 XP_016857145.1 4 - 5 557 89
XM_017001657.1 XP_016857146.1 3 - 4 345 89
XM_017001658.1 XP_016857147.1 4 - 5 488 89
XM_017001659.1 XP_016857148.1 2 - 3 378 89
XM_017001660.1 XP_016857149.1 2 - 3 281 89
XM_017001672.1 XP_016857161.1 2 - 3 378 89
XM_017001675.1 XP_016857164.1 2 - 3 378 89
XM_017001679.1 XP_016857168.1 3 - 4 490 89
GenBank mRNA AK090568.1 - 4 - 5 479 89
AK315649.1 - 3 - 4 406 89
BC014947.1 - 3 - 4 406 89
BX647626.1 - 3 - 4 493 89
DA072156.1 - 3 - 4 424 89
DA969344.1 - 2 - 3 306 89

View Full Product Details