The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181699.2 | NP_859050.1 | 13 - 14 | 1782 | 151 |
NM_181700.1 | NP_859051.1 | 11 - 12 | 1590 | 151 | |
XM_017017958.1 | XP_016873447.1 | 13 - 14 | 1726 | 151 | |
XM_017017959.1 | XP_016873448.1 | 13 - 14 | 1726 | 151 | |
XM_017017960.1 | XP_016873449.1 | 12 - 13 | 1591 | 151 | |
GenBank mRNA | AK292275.1 | - | 13 - 14 | 1702 | 151 |
AK301705.1 | - | 11 - 12 | 1531 | 151 | |
BC027596.2 | - | 13 - 14 | 1719 | 151 |