The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303012.1 | NP_001289941.1 | 16 - 17 | 2300 | 123 |
NM_001303013.1 | NP_001289942.1 | 16 - 17 | 3486 | 123 | |
NM_014638.3 | NP_055453.2 | 16 - 17 | 2494 | 123 | |
XM_011542455.2 | XP_011540757.1 | 17 - 18 | 2536 | 123 | |
XM_011542456.2 | XP_011540758.1 | 17 - 18 | 2493 | 123 | |
XM_017002870.1 | XP_016858359.1 | 17 - 18 | 2525 | 123 | |
XM_017002871.1 | XP_016858360.1 | 17 - 18 | 2496 | 123 | |
XM_017002872.1 | XP_016858361.1 | 17 - 18 | 2525 | 123 | |
XM_017002874.1 | XP_016858363.1 | 17 - 18 | 2525 | 123 | |
GenBank mRNA | AB007919.2 | - | 16 - 17 | 2300 | 123 |
AK074149.1 | - | 5 - 6 | 556 | 123 | |
AK122589.1 | - | 14 - 15 | 2593 | 123 | |
BC019679.1 | - | 2 - 3 | 122 | 123 | |
BC128207.1 | - | 14 - 15 | 1876 | 123 | |
BC144620.1 | - | 16 - 17 | 3478 | 123 | |
DQ176850.1 | - | 16 - 17 | 2494 | 123 |