The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002439.4 | NP_002430.3 | 3 - 4 | 663 | 140 |
GenBank mRNA | AI817671.1 | - | 3 - 4 | 140 | |
AK289856.1 | - | 4 - 5 | 632 | 140 | |
BC004177.2 | - | 4 - 5 | 604 | 140 | |
BC011817.2 | - | 4 - 5 | 604 | 140 | |
BC017273.2 | - | 3 - 4 | 586 | 140 | |
BC098436.1 | - | 4 - 5 | 604 | 140 | |
BC130434.1 | - | 3 - 4 | 663 | 140 | |
BC130436.1 | - | 3 - 4 | 663 | 140 | |
J04810.1 | - | 3 - 4 | 644 | 140 | |
U61981.1 | - | 4 - 5 | 572 | 140 |