The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002439.4 | NP_002430.3 | 4 - 5 | 874 | 149 |
GenBank mRNA | AK289856.1 | - | 5 - 6 | 843 | 149 |
BC004177.2 | - | 5 - 6 | 815 | 149 | |
BC011817.2 | - | 5 - 6 | 815 | 149 | |
BC017273.2 | - | 4 - 5 | 797 | 149 | |
BC098436.1 | - | 5 - 6 | 815 | 149 | |
BC130434.1 | - | 4 - 5 | 874 | 149 | |
BC130436.1 | - | 4 - 5 | 874 | 149 | |
J04810.1 | - | 4 - 5 | 855 | 149 | |
U61981.1 | - | 5 - 6 | 783 | 149 |