The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002439.4 | NP_002430.3 | 6 - 7 | 1108 | 87 |
GenBank mRNA | AK289856.1 | - | 7 - 8 | 1077 | 87 |
BC004177.2 | - | 7 - 8 | 1049 | 87 | |
BC011817.2 | - | 7 - 8 | 1049 | 87 | |
BC017273.2 | - | 6 - 7 | 1031 | 87 | |
BC098436.1 | - | 7 - 8 | 1049 | 87 | |
BC130434.1 | - | 6 - 7 | 1108 | 87 | |
BC130436.1 | - | 6 - 7 | 1108 | 87 | |
J04810.1 | - | 6 - 7 | 1089 | 87 | |
U61981.1 | - | 7 - 8 | 1017 | 87 |