Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TBC1D1
- Entrez Gene ID
-
23216
- Gene Name
- TBC1 domain family member 1
- Gene Aliases
-
TBC, TBC1
- Chromosome Location
-
Chr.4: 37891084 - 38139175 on Build GRCh38
View Full Product Details