Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AP4E1
Entrez Gene ID
23431
Gene Name
adaptor related protein complex 4 epsilon 1 subunit
Gene Aliases
CPSQ4, SPG51, STUT1
Chromosome Location
Chr.15: 50908569 - 51005900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252127.1 NP_001239056.1 10 - 11 1307 170
NM_007347.4 NP_031373.2 10 - 11 1284 170
XM_005254264.3 XP_005254321.1 10 - 11 1102 170
XM_006720447.3 XP_006720510.1 10 - 11 1462 170
XM_017022042.1 XP_016877531.1 10 - 11 1317 170
GenBank mRNA AB030653.1 - 10 - 11 1207 170
AF155156.2 - 10 - 11 1210 170
AK297290.1 - 10 - 11 1252 170
BC126308.1 - 10 - 11 1283 170
BC130466.1 - 10 - 11 1283 170
CR749604.1 - 10 - 11 1184 170

View Full Product Details