Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AP4E1
Entrez Gene ID
23431
Gene Name
adaptor related protein complex 4 epsilon 1 subunit
Gene Aliases
CPSQ4, SPG51, STUT1
Chromosome Location
Chr.15: 50908569 - 51005900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252127.1 NP_001239056.1 6 - 7 834 82
NM_007347.4 NP_031373.2 6 - 7 811 82
XM_005254264.3 XP_005254321.1 6 - 7 629 82
XM_006720447.3 XP_006720510.1 6 - 7 989 82
XM_017022042.1 XP_016877531.1 6 - 7 756 82
GenBank mRNA AB030653.1 - 6 - 7 734 82
AF155156.2 - 6 - 7 743 82
AK297290.1 - 6 - 7 811 82
BC126308.1 - 6 - 7 810 82
BC130466.1 - 6 - 7 810 82
CR749604.1 - 6 - 7 711 82

View Full Product Details