The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303143.1 | NP_001290072.1 | 2 - 3 | 379 | 100 |
GenBank mRNA | AK056134.1 | - | 1 - 2 | 596 | 100 |
AK293948.1 | - | 1 - 2 | 596 | 100 | |
BC003648.2 | - | 2 - 3 | 379 | 100 |