The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286076.1 | NP_001273005.1 | 1 - 2 | 357 | 111 |
NM_002946.4 | NP_002937.1 | 1 - 2 | 357 | 111 | |
GenBank mRNA | AK298759.1 | - | 1 - 2 | 135 | 111 |
AK300691.1 | - | 1 - 2 | 99 | 111 | |
AK312956.1 | - | 1 - 2 | 121 | 111 | |
BC001630.1 | - | 1 - 2 | 94 | 111 | |
BC012157.1 | - | 1 - 2 | 146 | 111 | |
BC021257.2 | - | 1 - 2 | 285 | 111 | |
DB025488.1 | - | 1 - 2 | 357 | 111 | |
J05249.1 | - | 1 - 2 | 80 | 111 |