The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001522.2 | NP_001513.2 | 5 - 6 | 1751 | 145 |
GenBank mRNA | BC144241.1 | - | 4 - 5 | 1546 | 145 |
L37378.1 | - | 5 - 6 | 1774 | 145 |