The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001522.2 | NP_001513.2 | 7 - 8 | 1973 | 125 |
GenBank mRNA | BC144241.1 | - | 6 - 7 | 1768 | 125 |
L37378.1 | - | 7 - 8 | 1996 | 125 |