The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000228.2 | NP_000219.2 | 10 - 11 | 1280 | 91 |
NM_001017402.1 | NP_001017402.1 | 9 - 10 | 1526 | 91 | |
NM_001127641.1 | NP_001121113.1 | 10 - 11 | 1254 | 91 | |
XM_005273124.4 | XP_005273181.1 | 10 - 11 | 1462 | 91 | |
XM_017001272.1 | XP_016856761.1 | 9 - 10 | 1690 | 91 | |
GenBank mRNA | AK296851.1 | - | 10 - 11 | 1230 | 91 |
AY035783.1 | - | 9 - 10 | 1136 | 91 | |
BC075838.1 | - | 9 - 10 | 1311 | 91 | |
D37766.1 | - | 10 - 11 | 1217 | 91 | |
KU178159.1 | - | 9 - 10 | 1136 | 91 | |
L25541.1 | - | 10 - 11 | 1253 | 91 |