Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMG20B
Entrez Gene ID
10362
Gene Name
high mobility group 20B
Gene Aliases
BRAF25, BRAF35, HMGX2, HMGXB2, PP7706, SMARCE1r, SOXL, pp8857
Chromosome Location
Chr.19: 3572938 - 3579083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006339.2 NP_006330.2 5 - 6 543 75
GenBank mRNA AF072165.1 - 2 - 3 449 75
AF072836.1 - 2 - 3 187 75
AF146223.1 - 4 - 5 485 75
AF288679.1 - 4 - 5 475 75
AF331191.1 - 4 - 5 475 75
AK090733.1 - 5 - 6 437 75
AK289500.1 - 5 - 6 531 75
AK314201.1 - 5 - 6 517 75
AL355691.1 - 3 - 4 441 75
AL355703.1 - 4 - 5 487 75
AL355709.1 - 4 - 5 485 75
BC002552.2 - 5 - 6 533 75
BC003505.1 - 5 - 6 531 75
BC004408.1 - 4 - 5 481 75
BC021585.1 - 3 - 4 445 75
CR456754.1 - 2 - 3 169 75

View Full Product Details