Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMG20B
Entrez Gene ID
10362
Gene Name
high mobility group 20B
Gene Aliases
BRAF25, BRAF35, HMGX2, HMGXB2, PP7706, SMARCE1r, SOXL, pp8857
Chromosome Location
Chr.19: 3572938 - 3579083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006339.2 NP_006330.2 9 - 10 1011 60
XM_017026144.1 XP_016881633.1 9 - 10 1015 60
GenBank mRNA AF072165.1 - 6 - 7 917 60
AF072836.1 - 6 - 7 656 60
AF146223.1 - 8 - 9 953 60
AF288679.1 - 8 - 9 943 60
AF318366.1 - 7 - 8 1833 60
AK090733.1 - 9 - 10 905 60
AK129638.1 - 4 - 5 1080 60
AK289500.1 - 9 - 10 999 60
AK314201.1 - 9 - 10 985 60
AL355691.1 - 7 - 8 909 60
AL355702.1 - 2 - 3 196 60
AL355703.1 - 8 - 9 955 60
AL355709.1 - 8 - 9 953 60
BC002552.2 - 9 - 10 1001 60
BC003505.1 - 9 - 10 999 60
BC004408.1 - 8 - 9 949 60
BC021585.1 - 7 - 8 913 60
BC134413.1 - 2 - 3 350 60

View Full Product Details