The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242307.1 | NP_001229236.1 | 13 - 14 | 1549 | 90 |
NM_001242309.1 | NP_001229238.1 | 12 - 13 | 1446 | 90 | |
NM_014889.3 | NP_055704.2 | 13 - 14 | 1549 | 90 | |
XM_005252345.3 | XP_005252402.1 | 13 - 14 | 1549 | 90 | |
XM_017015470.1 | XP_016870959.1 | 13 - 14 | 1617 | 90 | |
XM_017015471.1 | XP_016870960.1 | 13 - 14 | 1617 | 90 | |
XM_017015472.1 | XP_016870961.1 | 13 - 14 | 1446 | 90 | |
XM_017015473.1 | XP_016870962.1 | 13 - 14 | 1446 | 90 | |
GenBank mRNA | AB029027.2 | - | 3 - 4 | 278 | 90 |
AF061243.1 | - | 13 - 14 | 1493 | 90 | |
AK002061.1 | - | 13 - 14 | 1517 | 90 | |
AK023476.1 | - | 6 - 7 | 664 | 90 | |
AK225084.1 | - | 13 - 14 | 1487 | 90 | |
AK294865.1 | - | 12 - 13 | 1462 | 90 | |
AK299464.1 | - | 12 - 13 | 1295 | 90 | |
AK303406.1 | - | 12 - 13 | 1387 | 90 | |
AK315117.1 | - | 13 - 14 | 1487 | 90 | |
BC001150.1 | - | 7 - 8 | 849 | 90 | |
BC005025.1 | - | 13 - 14 | 1484 | 90 | |
BC045577.1 | - | 13 - 14 | 1959 | 90 | |
BC095422.1 | - | 13 - 14 | 1490 | 90 | |
BC111987.1 | - | 13 - 14 | 1505 | 90 | |
BC113369.1 | - | 13 - 14 | 1505 | 90 | |
BX649017.1 | - | 13 - 14 | 1491 | 90 | |
CR749279.1 | - | 11 - 12 | 1225 | 90 |