The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242307.1 | NP_001229236.1 | 15 - 16 | 1805 | 67 |
NM_001242309.1 | NP_001229238.1 | 14 - 15 | 1702 | 67 | |
NM_014889.3 | NP_055704.2 | 15 - 16 | 1805 | 67 | |
XM_005252345.3 | XP_005252402.1 | 15 - 16 | 1805 | 67 | |
XM_017015470.1 | XP_016870959.1 | 15 - 16 | 1873 | 67 | |
XM_017015471.1 | XP_016870960.1 | 15 - 16 | 1873 | 67 | |
XM_017015472.1 | XP_016870961.1 | 15 - 16 | 1702 | 67 | |
XM_017015473.1 | XP_016870962.1 | 15 - 16 | 1702 | 67 | |
GenBank mRNA | AB029027.2 | - | 5 - 6 | 534 | 67 |
AF061243.1 | - | 15 - 16 | 1749 | 67 | |
AK002061.1 | - | 15 - 16 | 1773 | 67 | |
AK023476.1 | - | 8 - 9 | 920 | 67 | |
AK225084.1 | - | 15 - 16 | 1743 | 67 | |
AK294865.1 | - | 14 - 15 | 1718 | 67 | |
AK299464.1 | - | 14 - 15 | 1551 | 67 | |
AK303406.1 | - | 14 - 15 | 1643 | 67 | |
AK315117.1 | - | 15 - 16 | 1743 | 67 | |
BC001150.1 | - | 9 - 10 | 1105 | 67 | |
BC005025.1 | - | 15 - 16 | 1740 | 67 | |
BC045577.1 | - | 15 - 16 | 2215 | 67 | |
BC095422.1 | - | 15 - 16 | 1746 | 67 | |
BC111987.1 | - | 15 - 16 | 1761 | 67 | |
BC113369.1 | - | 15 - 16 | 1761 | 67 | |
BX649017.1 | - | 15 - 16 | 1747 | 67 | |
CR749279.1 | - | 13 - 14 | 1481 | 67 |