The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003285.2 | NP_003276.3 | 5 - 6 | 1798 | 78 |
XM_011509949.2 | XP_011508251.1 | 4 - 5 | 1407 | 78 | |
XM_017002218.1 | XP_016857707.1 | 6 - 7 | 2235 | 78 | |
XM_017002219.1 | XP_016857708.1 | 5 - 6 | 2163 | 78 | |
GenBank mRNA | BC129830.1 | - | 3 - 4 | 1121 | 78 |
X98085.1 | - | 4 - 5 | 1360 | 78 | |
Z67996.1 | - | 3 - 4 | 1324 | 78 |