The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004963.3 | NP_004954.2 | 15 - 16 | 1847 | 101 |
XM_011520631.2 | XP_011518933.1 | 15 - 16 | 1745 | 101 | |
GenBank mRNA | BC136544.1 | - | 15 - 16 | 1756 | 101 |
BC136545.1 | - | 15 - 16 | 1756 | 101 | |
M73489.1 | - | 15 - 16 | 1758 | 101 |