The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004963.3 | NP_004954.2 | 24 - 25 | 3009 | 61 |
XM_011520631.2 | XP_011518933.1 | 24 - 25 | 2907 | 61 | |
GenBank mRNA | BC136544.1 | - | 24 - 25 | 2918 | 61 |
BC136545.1 | - | 24 - 25 | 2918 | 61 | |
M73489.1 | - | 24 - 25 | 2920 | 61 | |
S57551.1 | - | 24 - 25 | 2989 | 61 |