The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004963.3 | NP_004954.2 | 7 - 8 | 1081 | 104 |
XM_011520631.2 | XP_011518933.1 | 7 - 8 | 979 | 104 | |
GenBank mRNA | AK310034.1 | - | 7 - 8 | 1071 | 104 |
BC136544.1 | - | 7 - 8 | 990 | 104 | |
BC136545.1 | - | 7 - 8 | 990 | 104 | |
M73489.1 | - | 7 - 8 | 992 | 104 |