Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C12orf4
Entrez Gene ID
57102
Gene Name
chromosome 12 open reading frame 4
Gene Aliases
-
Chromosome Location
Chr.12: 4487730 - 4538508 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304811.1 NP_001291740.1 13 - 14 1678 123
NM_020374.3 NP_065107.1 13 - 14 1686 123
XM_005253716.1 XP_005253773.1 12 - 13 1557 123
GenBank mRNA AJ272205.1 - 13 - 14 1629 123
AK024811.1 - 5 - 6 527 123
AK074479.1 - 13 - 14 1686 123
AL834377.1 - 13 - 14 1649 123
BC016908.1 - 13 - 14 1629 123
BX641128.1 - 13 - 14 1678 123

View Full Product Details