Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C4orf33
Entrez Gene ID
132321
Gene Name
chromosome 4 open reading frame 33
Gene Aliases
-
Chromosome Location
Chr.4: 129093674 - 129112688 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099783.1 NP_001093253.1 3 - 4 340 148
NM_173487.2 NP_775758.2 3 - 4 612 148
XM_005262734.4 XP_005262791.1 3 - 4 607 148
XM_017007722.1 XP_016863211.1 3 - 4 1669 148
XM_017007723.1 XP_016863212.1 3 - 4 1419 148
XM_017007724.1 XP_016863213.1 3 - 4 595 148
GenBank mRNA AK091022.1 - 3 - 4 601 148
AK309229.1 - 3 - 4 586 148
BC032582.1 - 3 - 4 543 148
BQ549846.1 - 3 - 4 371 148
BX538164.1 - 3 - 4 553 148

View Full Product Details