The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291807.1 | NP_001278736.1 | 10 - 11 | 1138 | 64 |
NM_004460.3 | NP_004451.2 | 11 - 12 | 1213 | 64 | |
XM_011510796.2 | XP_011509098.1 | 10 - 11 | 1293 | 64 | |
XM_011510797.2 | XP_011509099.1 | 11 - 12 | 1154 | 64 | |
XM_017003585.1 | XP_016859074.1 | 11 - 12 | 1154 | 64 | |
GenBank mRNA | AK297118.1 | - | 10 - 11 | 1061 | 64 |
AK309260.1 | - | 10 - 11 | 994 | 64 | |
BC026250.1 | - | 11 - 12 | 1046 | 64 | |
KU257678.1 | - | 9 - 10 | 924 | 64 | |
U09278.1 | - | 11 - 12 | 1213 | 64 | |
U76833.1 | - | 11 - 12 | 1053 | 64 |