The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291807.1 | NP_001278736.1 | 1 - 2 | 215 | 91 |
NM_004460.3 | NP_004451.2 | 1 - 2 | 215 | 91 | |
XM_011510797.2 | XP_011509099.1 | 1 - 2 | 156 | 91 | |
XM_017003585.1 | XP_016859074.1 | 1 - 2 | 156 | 91 | |
GenBank mRNA | AK297118.1 | - | 1 - 2 | 138 | 91 |
AK309260.1 | - | 1 - 2 | 95 | 91 | |
AK315448.1 | - | 1 - 2 | 85 | 91 | |
U09278.1 | - | 1 - 2 | 215 | 91 |