The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039580.1 | NP_001034669.1 | 3 - 4 | 429 | 99 |
XM_006714306.3 | XP_006714369.1 | 3 - 4 | 577 | 99 | |
XM_011532253.1 | XP_011530555.1 | 3 - 4 | 632 | 99 | |
XM_011532254.2 | XP_011530556.1 | 3 - 4 | 541 | 99 | |
XM_011532255.2 | XP_011530557.1 | 2 - 3 | 280 | 99 | |
XM_017008616.1 | XP_016864105.1 | 3 - 4 | 632 | 99 | |
XM_017008617.1 | XP_016864106.1 | 3 - 4 | 449 | 99 | |
XM_017008618.1 | XP_016864107.1 | 3 - 4 | 407 | 99 | |
GenBank mRNA | AK024812.1 | - | 3 - 4 | 429 | 99 |
AK301075.1 | - | 3 - 4 | 296 | 99 | |
AY690636.1 | - | 3 - 4 | 429 | 99 | |
BC022852.1 | - | 3 - 4 | 257 | 99 | |
BC056683.1 | - | 2 - 3 | 213 | 99 | |
BC128540.1 | - | 3 - 4 | 344 | 99 | |
BC128541.1 | - | 3 - 4 | 344 | 99 | |
BC146864.1 | - | 3 - 4 | 341 | 99 |