The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166373.1 | NP_001159845.1 | 6 - 7 | 604 | 70 |
NM_017923.3 | NP_060393.1 | 2 - 3 | 491 | 70 | |
XM_011532055.2 | XP_011530357.1 | 6 - 7 | 1309 | 70 | |
XM_017008334.1 | XP_016863823.1 | 5 - 6 | 531 | 70 | |
XM_017008335.1 | XP_016863824.1 | 6 - 7 | 910 | 70 | |
XM_017008336.1 | XP_016863825.1 | 6 - 7 | 776 | 70 | |
GenBank mRNA | AK000675.1 | - | 2 - 3 | 380 | 70 |
AK289711.1 | - | 6 - 7 | 604 | 70 | |
AL713759.1 | - | 6 - 7 | 1399 | 70 | |
AV716666.2 | - | 2 - 3 | 490 | 70 |