The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101800.2 | NP_001095270.1 | 3 - 4 | 695 | 89 |
NM_016603.3 | NP_057687.2 | 3 - 4 | 695 | 89 | |
XM_005272007.1 | XP_005272064.1 | 3 - 4 | 695 | 89 | |
XM_006714647.1 | XP_006714710.1 | 3 - 4 | 695 | 89 | |
XM_006714648.1 | XP_006714711.1 | 3 - 4 | 447 | 89 | |
XM_011543448.1 | XP_011541750.1 | 4 - 5 | 488 | 89 | |
XM_011543449.1 | XP_011541751.1 | 4 - 5 | 792 | 89 | |
XM_011543450.1 | XP_011541752.1 | 3 - 4 | 404 | 89 | |
XM_017009549.1 | XP_016865038.1 | 3 - 4 | 720 | 89 | |
XM_017009550.1 | XP_016865039.1 | 3 - 4 | 404 | 89 | |
GenBank mRNA | AF251038.1 | - | 3 - 4 | 614 | 89 |
AK314224.1 | - | 3 - 4 | 671 | 89 | |
BC048965.1 | - | 3 - 4 | 667 | 89 | |
BC056887.1 | - | 2 - 3 | 270 | 89 | |
BU183000.1 | - | 2 - 3 | 269 | 89 |