Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM13B
Entrez Gene ID
51306
Gene Name
family with sequence similarity 13 member B
Gene Aliases
ARHGAP49, C5orf5, FAM13B1, KHCHP, N61
Chromosome Location
Chr.5: 137937909 - 138051961 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101800.2 NP_001095270.1 4 - 5 895 103
NM_001101801.2 NP_001095271.1 3 - 4 703 103
NM_016603.3 NP_057687.2 4 - 5 895 103
XM_005272007.1 XP_005272064.1 4 - 5 895 103
XM_006714647.1 XP_006714710.1 4 - 5 895 103
XM_006714648.1 XP_006714711.1 4 - 5 647 103
XM_006714649.1 XP_006714712.1 3 - 4 703 103
XM_011543448.1 XP_011541750.1 5 - 6 688 103
XM_011543449.1 XP_011541751.1 5 - 6 992 103
XM_011543450.1 XP_011541752.1 4 - 5 604 103
XM_017009549.1 XP_016865038.1 4 - 5 920 103
XM_017009550.1 XP_016865039.1 4 - 5 604 103
XM_017009551.1 XP_016865040.1 3 - 4 703 103
GenBank mRNA AF251038.1 - 4 - 5 814 103
AK314224.1 - 4 - 5 871 103
BC041335.1 - 3 - 4 681 103
BC048965.1 - 4 - 5 867 103
BC056887.1 - 3 - 4 470 103
BU183000.1 - 3 - 4 469 103

View Full Product Details