Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNF8
Entrez Gene ID
11267
Gene Name
SNF8, ESCRT-II complex subunit
Gene Aliases
Dot3, EAP30, VPS22
Chromosome Location
Chr.17: 48930096 - 48944842 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317192.1 NP_001304121.1 4 - 5 456 77
NM_001317193.1 NP_001304122.1 3 - 4 405 77
NM_007241.3 NP_009172.2 4 - 5 456 77
NR_133679.1 4 - 5 456 77
GenBank mRNA AF156102.1 - 4 - 5 405 77
BC008976.1 - 4 - 5 406 77
BC038830.1 - 4 - 5 430 77
BE818533.1 - 3 - 4 259 77
BG248541.1 - 4 - 5 445 77
BP342490.1 - 4 - 5 456 77

View Full Product Details