The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142351.1 | NP_001135823.1 | 5 - 6 | 67 | |
NM_001322362.1 | NP_001309291.1 | 5 - 6 | 67 | ||
NM_032528.2 | NP_115917.1 | 5 - 6 | 67 | ||
NR_136313.1 | 4 - 5 | 67 | |||
XM_006712802.1 | XP_006712865.1 | 5 - 6 | 67 | ||
XM_011511999.2 | XP_011510301.1 | 5 - 6 | 67 | ||
GenBank mRNA | AB058780.1 | - | 5 - 6 | 1437 | 67 |
AB059555.1 | - | 5 - 6 | 1493 | 67 | |
AJ512141.1 | - | 3 - 4 | 378 | 67 |