The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003298.4 | NP_003289.2 | 2 - 3 | 436 | 63 |
XM_011534058.2 | XP_011532360.1 | 5 - 6 | 471 | 63 | |
XM_011534059.2 | XP_011532361.1 | 5 - 6 | 1377 | 63 | |
XM_011534061.2 | XP_011532363.1 | 3 - 4 | 325 | 63 | |
XM_011534064.2 | XP_011532366.1 | 4 - 5 | 574 | 63 | |
XM_011534065.2 | XP_011532367.1 | 3 - 4 | 284 | 63 | |
XM_011534066.2 | XP_011532368.1 | 2 - 3 | 1087 | 63 | |
XM_017007118.1 | XP_016862607.1 | 3 - 4 | 553 | 63 | |
XM_017007119.1 | XP_016862608.1 | 3 - 4 | 213 | 63 | |
GenBank mRNA | BC051670.1 | - | 2 - 3 | 209 | 63 |
HQ995528.1 | - | 1 - 2 | 74 | 63 | |
L27586.1 | - | 2 - 3 | 296 | 63 |